Ledverlichtingssystemen, EasyLED
Leverancier: SCHOTT AG Lighting and imaging
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
600101 600121 600121-SETC 600121-SETSA
630-2662EA 453 EUR
630-2662 630-2663 630-3491 630-3492
Ledverlichtingssystemen, EasyLED
Lampen LED-verlichting
The SCHOTT EasyLED portfolio for stereo microscopy delivers precise LED light for a wide variety of applications, such as laboratory and diagnostics, industrial applications, education, or forensic science. The series offers robust design and easy handling in a compact device, with a range of accessories for specific requirements.
  • Powerful luminance; Intense light output for specialised tasks
  • Maintenance-free LED lifetime of 50000 hours
  • Color temperature: approximately 5600 K
  • Low power consumption; smart engineering leads to optimized energy use
  • Compatible with all common OEMs
  • Mounting adapter is available for all common microscope stands, making the EasyLED suitable for a wide range of systems
  • Ergonomic design; Integrated controller enables simple and intuitive switching between illumination modes
Spotlight Plus
Maximale lichtflux van 140 lm per spot
Dubbele Spotlight Plus met schakelfunctie: ledkop 1 AAN, ledkop 2 A, beide ledkoppen AAN, beide ledkoppen UIT
Spotlight Plus-adapters voor alle voeten en losstaande versie beschikbaar
Ringlight: Maximale lichtflux van 90 klx bij een vrije werkafstand (FWD) van 75 mm met 45 led's met een hoge helderheid, eenvoudig aan te brengen op diverse microscoopobjectieven (Ø 66 mm/adapters verkrijgbaar) en een breed bereik aan werkafstanden (55-135 mm).
Spot lights: EasyLED spot lights are available in two different versions; an individual spot light with a single light head (spot light plus single), and a double spot light with two light heads (spot light plus double). In addition, the two light heads of the spot light plus double can be used individually.
Tafel voor doorvallend licht: Met zeer uniforme achtergrondverlichting (Ø 50 mm), gemakkelijk uit te breiden naar voet voor invallend licht (Ø 84 mm/adapters verkrijgbaar) en maximale luminantie tot 12.000 cd/m².
The new SCHOTT® tubular diffuser for LED ring lights avoid interfering reflections when viewing the surfaces of non-transparent samples with incident light. Allows free access to the sample compared to dome lights and offers more diffuse light.
Certificaten: Power supply unit, UL, PSE, EMC-class B.
Informatie over toebehoren: SCHOTT offers a variety of mechanical and optical accessories to increase the flexibility of the EasyLED series. Accessories include polarisation filters. SCHOTT ensures that all accessories offer simple-to-use operation and a long lifetime.
Bestelinformatie: Spotlights are supplied without mounting adapters, except in the Spotlight sets - see mounting brackets.