Single channel pipettes, electronic, variable volume, Ovation®
Leverancier: VISTALAB TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1057-0002 1057-0010 1057-0020 1057-0100 1057-0200 1057-1000
613-4921EA 1320 EUR
613-4921 613-4922 613-4923 613-4924 613-4925 613-4926
Single channel pipettes, electronic, variable volume, Ovation®
Pipetten Micropipetten
Manually operated pipette with quick set digital volume adjustment.
  • Easy to program and calibrate for different environmental conditions and liquids
  • Pipettes stand flat on the bench so no pipette stands are required
  • Motor-driven volume adjustment for ease of change
  • One-way disposable nozzle with built-in filter to prevent pipette contamination
  • Colour-coded for easy volume recognition
* Certified DNase/RNase free
Bestelinformatie: Supplied with a lithium ion battery (giving approximately six months use).