Peristaltic pumps, triple-channel
Leverancier: WATSON MARLOW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
040.181D.M30 040.1P1D.M30
224-0369EA 0 EUR
224-0369 181-2214
Peristaltic pumps, triple-channel
Pompen Peristaltische pompen
401U/DM3 three channel pump accepts double segment manifold tubing for microlitre flows.
  • 401U/DM3: triple–channel, flow rates up to 36 ml/min per channel
  • Pumpheads have adjustable occlusion for precision flow rates
  • Very compact design, single potentiometer for rotation direction, speed and start/stop
  • Analogue speed control and external control via a 15–pin Sub–D connector
  • Reversible rotation
Control options: analogue signals 4 to 20 mA or 0 to 10 V, TTL signal (SPS compatible), start/stop and rotation direction. The precision–engineered four–rollerMore Product Information