Accessories for Spectroquant® NOVA
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.09769.0001 1.14723.0001 1.14964.0001 1.09749.0001
1.09769.0001EA 0 EUR
1.09769.0001 1.14723.0001 1.14964.0001 1.09749.0001
Accessories for Spectroquant® NOVA
Spectrofotometers en fotometers UV/Vis spectrofotometers
To document the analysis values, either a standard computer or a printer can be connected.
  • The Multi-Agate II software can control data acquisition or simply record recorded analysis data
  • The data records are archived here as raw data or imported into further programs such as Excel
Connectable photometers: SQ 118, SQ 200, SQ 300, Spectroquant® NOVA 30, Spectroquant® NOVA 60, Spectroquant® NOVA 400.