Stirrer Glands, PFTE or Brass, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ST4R ST4W
201-8732EA 35.3 EUR
201-8732 201-8733
Stirrer Glands, PFTE or Brass, Quickfit®
Roerders Roergereedschap
Stuffing box type glands with choice of brass or PTFE cones. Accepts 6 mm diameter stirrer shaft.
  • Provides a seal by means of adjustable compression of O rings and lubricated felt washers
  • 201-8730 and 201-8731 have integral drive pulleys