Stirrer Glands, Glass, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ST20/2 ST20/3 ST20/4
201-8727EA 197 EUR
201-8727 201-8728 BIBBST20/4
Stirrer Glands, Glass, Quickfit®
Roerders Roergereedschap
Parallel ground glass sleeve with cup to accept suitable lubricant for introducing stirrer paddles into Quickfit® flasks and vessels.
  • Provides vapour-tight seal between paddle and flask
  • Holds vacuum down to 13,3 Pa
Bestelinformatie: Supplied with screw caps, rubber rings and PTFE washers. Note the stirrer should not be run at speeds in excess of 700 min⁻¹.