Roerstaafgeleiders met linkse schroefdraad
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
06306004 06306204
LENZ06306004EA 294 EUR
LENZ06306004 LENZ06306204
Roerstaafgeleiders met linkse schroefdraad
Roerders Roergereedschap
DURAN®, borosilicate glass, PFTE/fibre glass pressure screw. The fluoroplastic bearing is in a borosilicate glass housing, and has adjustable PTFE/FPM seal. All parts are interchangeable.