Stirrer with paddles
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
06308001 06308003 06308005 06308007 06308009
LENZ06308001EA 86.1 EUR
LENZ06308001 441-0049 LENZ06308005 LENZ06308007 LENZ06308009
Stirrer with paddles
Roerders Roergereedschap
DURAN®, borosilicate glass shaft, PFTE paddles with ground and polished shaft.
Opgelet: * stirring Ø when blades extended