Digitale precisiethermometers, HACCP, TFX 410
Leverancier: EBRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1340-5418 1340-5419 1340-5410 1340-5415 1340-5417 1340-4169 1340-5416 1340-4174 1340-4183 1340-4184
620-0212EA 359 EUR
620-0212 620-0213 620-1473 620-1463 620-1459 620-1457 620-1458 EBRO1340-4174 EBRO1340-4183 EBRO1340-4184
Digitale precisiethermometers, HACCP, TFX 410
Thermometers Digitale thermometers
This handy and light thermometer is equipped with a large screen for easy reading. A selection of probes can be used with the thermometer for a wide range of applications.
  • Waterproof and dishwasher-proof
  • Cable and handle can withstand temperatures up to 200 °C
  • Easy to calibrate
  • High degree of accuracy
  • Automatic switch-off after two hours
Available with different PT1000 probes.
W×D×H: 54×22×109 mm
Weight: 90 g
Certificaten: DIN EN 13485.
Leveringsinformatie: Supplied with 3 V lithium battery for service life of about 5 years and a 3-point calibration certificate. 620-1463 without probe.