Secchi disc
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
707-0005EA 145 EUR
707-0005
Secchi disc
Turbiditeitsmeters
The disc is designed to measure water transparency in situ via the semi-quantitative method.
  • Disc slowly lowered in the water until the pattern is no longer visible
  • Depth of immersion is then evaluated
  • Measure related to the transparency
For use in oceans and lakes.
Ø = 200 mm
Certificaten: ISO 7027