Membraan-, druk- en vacuümpompen, LABOPORT®
Leverancier: KNF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N035AN.18 N86KN.18 N86KT.18 N816.3KT.18 N816.1.2KT.18 N035.3AT18IP20 N816.3KT.45.18
181-3233EA 756 EUR
181-3233 181-3281 181-3282 181-3135 181-3109 181-0344 181-3917
Membraan-, druk- en vacuümpompen, LABOPORT®
Pompen Membraanvacuümpompen
Single- or dual-headed, dry running diaphragm pumps for a wide variety of applications in laboratories. They transfer and evacuate without affecting the media, i.e. media remains uncontaminated. The pump includes a PTFE-coated diaphragm with patented tension optimisation, which allows the pump to be smaller and prolongs the service life of the diaphragm, it is ideal for aggressive/corrosive gases and vapors.
  • 100% oil-free transfer, evacuation and compression, so media remains uncontaminated
  • Version available for neutral to mildly aggressive gases and vapours
  • Maintenance-free, quiet running
  • Environmentally friendly; the pumps use no water and so produce no contaminated waste water
  • Resistant materials for pump head parts coming into contact with media
  • Flow rate up to 1,8 m³/h / Ultimate vacuum up to 15 mbar abs.
  • N 938.50 KT.18 ensures exceptionally fast evacuation by the parallel and in series connection of both pump heads
Application: Filtration, SPE, degassing, fluid aspiration.
Materiaal voor pompkoponderdelen die met media in contact komen
KN: PPS pompkop; EDPM membraan; FPM ventielen
KT: PPS pompkop; PTFE-coated membraan; FFPM ventielen
AN:aluminium pompkop; CR membraan; NBR ventielen
AT: aluminium pompkop; PTFE-coated membraan; FFPM ventielen
KT: PPS pompkop; PTFE-coated membraan; FFPM ventielen
AN:aluminium pompkop; CR membraan; NBR ventielen
AT: aluminium pompkop; PTFE-coated membraan; FFPM ventielen
Bestelinformatie: gelieve onze site www.vwr.com te raadplegen voor meer details over het volledige gamma LABOPORT pompen.
Opgelet: *Debiet bij atmosferische druk