Toebehoren voor vacuümschoepenpompen, RE/RZ
Leverancier: VACUUBRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20698000 20698003 20698004 20698006 20698007 20698017 20667052 22614149
181-1211EA 352 EUR
181-1211 181-1221 181-1224 181-1212 181-1213 181-1222 181-0481 VACU22614149
Toebehoren voor vacuümschoepenpompen, RE/RZ
Pompen Roterende vacuümschoepenpompen
Separator inlet side (AK)
Separators at the inlet protect pumps from particulates and droplets which may shorten service intervals and even reduce the lifetime and the operating performance of oil-sealed rotary vane pumps. Separators (AK) protect the vacuum pump at the inlet from particles and liquid droplets. Separators catchpot made of plastic (PMP) with good chemical resistance.
Note for RE/RZ 2.5: For rotary vane pumps RE 2.5 with article no. 2079715x and RZ 2.5 with article no. 2079812x oder separator 20698006. For rotary vane pumps RE 2.5 with article no. 2069715x and RZ 2.5 withMore Product Information