Systemen voor omgekeerde osmose, RO 10 / 20 / 30 / 40
Leverancier: ENVIROFALK PHARMAWATERSYSTEMS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1981-001 1981-002 1981-003 1981-004
172-4211EA 9070 EUR
172-4211 172-4212 172-4213 172-4214
Systemen voor omgekeerde osmose, RO 10 / 20 / 30 / 40
Waterzuiveringssystemen Puur- en ultrapuur-watersystemen
These compact units with two or three purification steps produce economically demineralised water directly from tap water without any pretreatment. Ideal for general Lab requirements, such as dishwashers, autoclaves, glass cleaning, simple analytics or pretreatment for ultrapure water systems.
  • Integrated pre filter, automatic flush system and reliable stainless steel pressure vessels
  • Two integrated conductivity meters for monitoring the whole system
  • RS 232 and centronics port allows a system monitoring according to GLP
  • Modular concept: Can be extended to meet increased pure water requirements