Schiff reagent for microscopical staining
Leverancier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
X900.1
X900.2
ROTHX900.1EA
33.9
EUR
ROTHX900.1
ROTHX900.2
Schiff reagent for microscopical staining
Schiff reagent
Staining reagent for detection of carbohydrates in tissue (PAS reaction).
Fuchsine sulphurous acid
Storage: At room temperature in the dark. After the bottle has been opened well-sealed at 4-8 °C. Discard the solution if it turns reddish.