Sigma-Aldrich® Bouin's solution
Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HT101128-4L
HT10132-1L
HT101128-4LEA
415
EUR
HT101128-4L
HT10132-1L
Sigma-Aldrich® Bouin's solution
Kleurstoffen
Microscopy and staining reagents
Bouin's solution is an excellent fixative for preserving soft and delicate structures. It is prepared with saturated picric acid, formaldehyde and acetic acid.
- Acetic acid 5%
- Formaldehyde 9%
- Picric acid 0,9%
Used as a mordant in various trichrome procedures.