Resazurin sodium salt
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
7-Sodiooxy-3H-phenoxazin-3-one 10-oxide, Sodium 7-oxido-3-oxo-3H-phenoxazine 10-oxide, Resazurin sodium, Resazurin
34226.01
34226.02
SERA34226.01EA
0
EUR
SERA34226.01
SERA34226.02
Resazurin sodium salt
Resazurin sodium salt
Formule:
C₁₂H₆NNaO₄ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00005036 CAS nummer: 62758-13-8 EINECS: 263-718-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...