Lakmoes
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0215526325 0215526380
ICNA0215526325EA 147 EUR
ICNA0215526325 ICNA0215526380
Lakmoes
Lakmoes
MDL Nummer: MFCD00131528
CAS nummer: 1393-92-6
EINECS: 215-739-6