β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form)
Leverancier: PanReac AppliChem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Nicotinamide adenine dinucleotide reduced, disodium salt (reduced form), NADH disodium salt, ((2R,3S,4R,5R)-5-(6-Aminopurin-9-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy-((((2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy)hydroxyphosphoryl)oxyphosphinic acid disodium salt
A1393.0001 A1393.0010
A1393.0001EA 76.6 EUR
A1393.0001 A1393.0010
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form)
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form)
Formule: C₂₁H₂₇N₇Na₂O₁₄P₂
MW: 709.41 g/mol
MDL Nummer: MFCD00036200
CAS nummer: 606-68-8
EINECS: 210-123-3