ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form)
Leverancier: PanReac AppliChem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: NADP monosodium salt, NADP Sodium, Nadide phosphate monosodium salt, Sodium (2R,3R,4R,5R)-2-(6-amino-9H-purin-9-yl)-5-((((((((2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)(hydroxy)phosphoryl)oxy)methyl)-4-hydroxytetrahydrofuran-3-yl hydrogen phosphate, Nicotinamide-adenine dinucleotide phosphate monosodium salt, NADP sodium salt, NADP-Na
A1394.0001 A1394.0005 A1394.9050
A1394.0001EA 198 EUR
A1394.0001 A1394.0005 A1394.9050
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form)
Formule: C₂₁H₂₇N₇NaO₁₇P₃
Smeltpunt: 175 to 178 °C
MDL Nummer: MFCD00036973
CAS nummer: 1184-16-3
EINECS: 214-664-6