di-Sodium β-glycerophosphate pentahydrate 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Sodium β-glycerophosphate pentahydrate, Disodium-β-glycerophosphate pentahydrate, Glycerol-2-phosphate, β-GP, di-Sodium-β-glycerophosphate pentahydrate, β-Glycerophosphoric acid disodium salt pentahydrate, β-glycerolphosphate
410990250 410991000 410995000
ACRO410990250EA 46.5 EUR
ACRO410990250 ACRO410991000 ACRO410995000
di-Sodium β-glycerophosphate pentahydrate 98%
di-Sodium β-glycerophosphate pentahydrate
Formule: C₃H₇Na₂O₆P·5H₂O
MW: 306.11 g/mol
Smeltpunt: 102…104 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002135
CAS nummer: 13408-09-8
EINECS: 212-464-3
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
Isomer ≤2 % alpha-Isomer