SSC 20X buffer, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
092810305 092810306 092810307
ICNA092810305EA 155 EUR
ICNA092810305 ICNA092810306 ICNA092810307
SSC 20X buffer, MP Biomedicals
Buffers Buffers voor elektroforese Wasbuffers
Role of a balanced salt solution in cell culture is multifaceted and can be divided into four principal functions:
  • Serves as an irrigating, transporting and diluting fluid while maintaining intra- and extracellular osmotic balance
  • Provides cells with water and certain bulk inorganic ions essential for normal cell metabolism
  • Combined with a carbohydrate, such as glucose, provides the principal energy source for cell metabolism
  • Provides a buffering system to maintain the medium within the physiological pH range (7,2 to 7,6)
Phosphate buffered saline tablets have been specifically developed for use in immunoassay procedures.