VWR®, Rekken voor cryodozen
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
479-1169EA 71.7 EUR
479-1169 479-1170
VWR®, Rekken voor cryodozen
Rekken Rekken voor cryogene dozen
Cardboard
  • Stable and long lasting
  • Lightweight - the 20 compartment rack weighs 1,1 kg without boxes
  • For cryoboxes 50 mm height
Economical, coated cardboard racks for upright freezers.