2-(Cyclohexylamino)ethanesulphonic acid (CHES)

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

17672.02 17672.03
SERA17672.02EA 0 EUR
SERA17672.02 SERA17672.03
2-(Cyclohexylamino)ethanesulphonic acid (CHES)
2-(Cyclohexylamino)ethanesulphonic acid (CHES)
Formule: C₈H₁₇NO₃S
Smeltpunt: 320 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003835
CAS nummer: 103-47-9
EINECS: 203-115-6
Merck Index: 13,02063

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR