Sodium tetrachloroaurate dihydrate ≥99.99% (metals basis) Au 49-50%, Premion®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Sodium tetrachloroaurate(III) dihydrate
12148.03
12148.06
12148.03EA
170
EUR
12148.03
12148.06
Sodium tetrachloroaurate dihydrate ≥99.99% (metals basis) Au 49-50%, Premion®
Sodium tetrachloroaurate dihydrate
Purity: 99.99%
CAS No.: 13874-02-7
Molecular Formula: AuCl4Na.2H2O
Formula Weight: 397.80 (361.77anhy)
Physical Form: Crystalline
Density: 0.8
Appearance: Yellow-orange
Melting Point: 100° dec.
Solubility: Soluble in water, alcohol, ether
Merck Reference: 14,8688
UN No.: 3260
MDL No.: MFCD00149162
Formule:
NaAuCl₄·2H₂O MW: 397.8 g/mol Smeltpunt: 100 °C (ontbinding) Dichtheid: 0.8 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00149162 CAS nummer: 13874-02-7 EINECS: 239-241-3 UN: 3260 ADR: 8 III Merck Index: 14,8688 |