Insulinespuiten, BD Micro-Fine™
Leverancier: EMBECTA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
324892 324891 324826 324825 324827 324824
EMBE324892EA 136 EUR
EMBE324892 EMBE324891 EMBE324826 EMBE324825 EMBE324827 EMBE324824
Insulinespuiten, BD Micro-Fine™
Spuiten Injectiespuiten, medisch
Micro-Fine™ is a single-use syringe for diabetes. It offers a more comfortable and convenient way to inject insulin with three different volumes.
  • Made from surgical stainless steel
  • Precision and comfort
  • Optimum shape and sharpness of the needle tip and bevel
  • Electro polished needle shaft
The white and orange protective cap conforms with sterility and a minimal dead space reduces the risk of air bubbles and prevents waste of insulin.