Centrifugebuisjes, Nunc™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
347708 347759 347856 347880 348224 337846
525-0056EA 455 EUR
525-0056 391-8505 391-8506 525-0057 391-8512 NUNC337846
Centrifugebuisjes, Nunc™
Buisjes Centrifugebuisjes
Polycarbonate or polystyrene.
  • Sterile
  • Good chemical resistance
  • Good temperature resistance
Conical, flat or round bottom.