Trechters met korte steel, PP
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
147010 147015 147020 147025 147030 147035
BRND147010EA 24.8 EUR
BRND147010 BRND147015 BRND147020 BRND147025 BRND147030 BRND147035
Trechters met korte steel, PP
Trechters Filtertrechters Filtertrechters
PP, translucent.
  • Excellent chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper
With smooth interior surfaces.