Flessen, brede hals, PPCO, economy, met schroefdop, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2187-0001 2187-0004 2187-0032
215-2243EA 85.1 EUR
215-2243 215-2248 215-2550
Flessen, brede hals, PPCO, economy, met schroefdop, Nalgene®
Flessen Kunststof flessen
PPCO, translucent.
  • Autoclavable
  • Economical and recyclable
  • Translucent for easy viewing of liquid levels
  • Leakproof when used with Nalgene closures
  • Manufactured from non-toxic materials
  • Very good chemical and temperature resistance
Light-duty, for general purpose applications where heavier bottles are not required.
Linerless caps for leakproof performance without a liner that can wrinkle, cause leaks and contaminate your solutions. Made from high-quality, laboratory-grade plastic materials for reliably low leachables and extractables.
Leveringsinformatie: Includes polypropylene screw closures.