Natrium, paraffin protect rods for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M-Clarity™ quality level = MQ200
8.22284.0250 8.22284.1000 8.22284.0100
8.22284.0250EA 131 EUR
8.22284.0250 8.22284.1000 8.22284.0100
Natrium, paraffin protect rods for synthesis, Sigma-Aldrich®
Natrium
Sodium rods (protective liquid: paraffin oil)
Formule: Na
MW: 22.99 g/mol
Kookpunt: 889 °C (1013 hPa)
Smeltpunt: 97.8
Dichtheid: 0.97 g/cm3 (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00085307
CAS nummer: 7440-23-5
EINECS: 231-132-9
UN: 1428
ADR: 4.3 I
Merck Index: 13,08641
Order Now
Specification Test Results
Content at Na ≥ 99.0 %