Beryllium ≥99.9% (metals basis), ultrathin foil, Thickness: 1 µm
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
40709.FF 40709.FI
40709.FFEA 0 EUR
40709.FF 40709.FI
Beryllium ≥99.9% (metals basis), ultrathin foil, Thickness: 1 µm
Beryllium
Purity: 99.9% CAS No.: 7440-41-7 Physical Form: On temporary copper support Odour: Odourless MDL No.: MFCD00134032
Alloy source, X-ray tube manufacture, neutron source
Formule: Be
Kookpunt: 2970 °C (1013 hPa)
Smeltpunt: 1278 °C
Dichtheid: 1,85 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134032
CAS nummer: 7440-41-7
EINECS: 231-150-7
Merck Index: 12,01201