Strontium ≥99% (metals basis), granules ≤ 19 mm (0.76 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Packaged under argon.
35789.06 35789.14 35789.22
35789.06EA 57.5 EUR
35789.06 35789.14 35789.22
Strontium ≥99% (metals basis), granules ≤ 19 mm (0.76 in)
Strontium
Purity: 99% CAS No.: 7440-24-6 Physical Form: Packaged under argon with thin oil layer Sensitivity: Air and Moisture Sensitive Appearance: White to pale yellow UN No.: 3208 MDL No.: MFCD00134060
Formule: Sr
MW: 87.62 g/mol
Kookpunt: 1151
Smeltpunt: 570…660 °C
Dichtheid: 2.38 g/cm³ (770 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134060
CAS nummer: 7440-24-6
EINECS: 231-133-4
UN: 3208
ADR: 4.3 II
Merck Index: 12,08994