Verzendverpakkingen
Leverancier: DAKLAPACK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
460177 460186 460144 460148 460168
216-0802EA 269 EUR
216-0802 216-0790 216-0800 216-0801 DAKL460168
Verzendverpakkingen
Mailer enveloppen en transportverpakkingen
Solid PP.
  • Transport blisters are designed for sending medical and biological samples
  • Can be sent as letter, saving significant postage costs
Used in combination with the liquid absorbent insert and SafetyBag, they meet the packaging requirements of most international postal services.