Cryodozen B40/D40
Leverancier: GLW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D40 D40B D40G D40R D40Y D40S DS40 D70 D70B D70G D70R D70Y D70S DS70 D90 D90B D90G D90R D90Y D90S DS90 D60 D60B D60G D60R D60S D60Y DS60
211-0525EA 5.7 EUR
211-0525 211-0526 211-0527 211-0528 211-0530 211-0529 479-1513 GLWAD70 GLWAD70B GLWAD70G GLWAD70R GLWAD70Y GLWAD70S GLWADS70 211-0531 211-0532 211-0533 211-0534 211-0536 211-0535 479-1566 GLWAD60 GLWAD60B GLWAD60G GLWAD60R GLWAD60S GLWAD60Y GLWADS60
Cryodozen B40/D40
Dozen Dozen voor cryobuisjes
Cryoboxes, PP, natural or coloured, without divider.
  • Autoclavable up to +121 °C and use in freezer down to –90 °C
  • The included clear lid offers a view of the contents without opening the cryobox
  • Without drainage holes in base for catching drips or store fluids (leakproof only at bottom up to 40 mm)
  • For light sensitive samples please choose the colour Black/black
The boxes comes without compartments, in addition they can be used with GLW dividers for individual configuration.They are suitable to store non standardised tubes, stone samples, small items and more. Inner dimension of the box: 124×124 mm, outer dimension of the box: 130×130 mm.
D40-series
Empty box, without divider. Bottom part of the box – height 45 mm / Lid - height 0 up to 10 mm
D60-series
Empty box, without divider. Bottom part of the box – height 45 mm / Lid - height 7 mm
D70-series
Empty box, without divider. Bottom part of the box – height 45 mm / Lid - height 30 up to 35 mm
D90-series
Empty box, without divider. Bottom part of the box – height 45 mm / Lid - height 45 up to 50 mm.