Flessen, smalle hals met schroefdop, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2204-0010 2204-0005 2204-0020
NALG2204-0010EA 45.4 EUR
NALG2204-0010 NALG2204-0005 NALG2204-0020
Flessen, smalle hals met schroefdop, Nalgene®
Flessen Kunststof flessen
PPCO, amber.
  • Protect light-sensitive liquids, reduce UV-light
  • Excellent chemical resistance to most acids, bases and alcohols.
  • Very good temperature resistance
  • Meet the U.S. Pharmacopoeia 23 requirements for light transmission
  • Autoclavable/Leakproof when used with Nalgene closures
  • Bottle and carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
These bottles are useful for storing chemicals and standards and for collecting and storing distilled water.
Linerless caps provide the ultimate in leakproof protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents.
4 and 8 L sizes feature a built-in shoulder loop on larger sizes for attaching identification tag.