Copper ≥99.998% (metals basis), foil, Puratronic® oxygen-free, Thickness: 0.5 mm (0.02 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
36395.FI 36395.GB
36395.FIEA 154 EUR
36395.FI 36395.GB
Copper ≥99.998% (metals basis), foil, Puratronic® oxygen-free, Thickness: 0.5 mm (0.02 in)
Copper
Purity: 99.998% CAS No.: 7440-50-8 Physical Form: ≈11.2g/50x50mm Odour: Odourless MDL No.: MFCD00010965
Formule: Cu
Kookpunt: 2595 °C (1013 hPa)
Smeltpunt: 1083 °C
Dichtheid: 8,94 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010965
CAS nummer: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545