Reageerbuisrekken
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
115510 115515 115520
BRND115510EA 133 EUR
BRND115510 BRND115515 BRND115520
Reageerbuisrekken
Rekken Tube Racks
Racks, made of PTFE.
  • Excellent chemical resistance
  • Temperature resistant from –200 to +280 °C
  • Non stick surfaces for easy cleaning
Robust and inert to virtually all chemical reagents with very good thermal resistance. Suitable for use in cryogenics, freezers, ovens.