Filtreertrechters
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
464003 464004 464713 464714 465423 465424 466133 466134
BRND464003EA 56 EUR
BRND464003 BRND464004 BRND464713 BRND464714 BRND465423 BRND465424 BRND466133 BRND466134
Filtreertrechters
Trechters Filtertrechters Filtertrechters
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filters disc.