Copper ≥99.9999% (metals basis), slug, Puratronic®, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42961.14 42961.22
42961.14EA 95.7 EUR
42961.14 42961.22
Copper ≥99.9999% (metals basis), slug, Puratronic®, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Copper
Purity: 99.9999% CAS No.: 7440-50-8 Physical Form: ≈3.6g/slug Odour: Odourless MDL No.: MFCD00010965
Formule: Cu
MW: 63.55 g/mol
Kookpunt: 2595
Smeltpunt: 1083
Dichtheid: 8.94 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010965
CAS nummer: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545