Copper ≥99% (metals basis), soluble -325 mesh
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
45073.30 45073.A1
45073.30EA 115 EUR
45073.30 45073.A1
Copper ≥99% (metals basis), soluble -325 mesh
Copper
Purity: 99% CAS No.: 7440-50-8 Molecular Formula: Cu Formula Weight: 63.55 Physical Form: Flake, average thickness 1-2 microns Density: 8.94 Melting Point: 1083° Boiling Point: 2562° UN No.: 3089 MDL No.: MFCD00010965
Formule: Cu
MW: 63.55 g/mol
Kookpunt: 2562 °C
Smeltpunt: 1083
Dichtheid: 8.94
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010965
CAS nummer: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1 III
Merck Index: 13,02545