Zink 99,999 Metals basis, foil, Puratronic®, Thickness: 2 mm (0.08 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
43504.FF 43504.FI
43504.FFEA 317 EUR
43504.FF 43504.FI
Zink 99,999 Metals basis, foil, Puratronic®, Thickness: 2 mm (0.08 in)
Zink
Purity: 99.999% CAS No.: 7440-66-6 Physical Form: ≈8.9g/25x25mm Odour: Odourless MDL No.: MFCD00011291
Formule: Zn
MW: 65.39 g/mol
Kookpunt: 908
Smeltpunt: 420
Dichtheid: 7.14 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011291
CAS nummer: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255
Order Now
Specificaties
Environmentally Preferable