Magnetische roerstaven, PTFE, ovaal
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
137300 137301 137302 137305 137307 137310 137312 137315 137320 137325
BRND137300EA 89.7 EUR
BRND137300 BRND137301 BRND137302 BRND137305 BRND137307 BRND137310 BRND137312 BRND137315 BRND137320 BRND137325
Magnetische roerstaven, PTFE, ovaal
Roerders Magnetische roerstaven
These oval shaped PTFE magnetic stirrer bars are ideal for vessels with round bottom such as round bottom flasks.