Zink 99.995% Trace metals basis, powder
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
194500050 194500500
ACRO194500050EA 130 EUR
ACRO194500050 ACRO194500500
Zink 99.995% Trace metals basis, powder
Zink
Formule: Zn
MW: 65.39 g/mol
Kookpunt: 908
Smeltpunt: 420 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011291
CAS nummer: 7440-66-6
EINECS: 231-175-3
UN: 1436
ADR: 4.3 II
Merck Index: 12,10255
Order Now
Specification Test Results
Appearance Grey Powder
Total trace metal impurities ≤50 ppm