Rhenium 99,97 Metals basis, foil, Thickness: 0.25 mm (0.01 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10309.FF 10309.FI
10309.FFEA 274 EUR
10309.FF 10309.FI
Rhenium 99,97 Metals basis, foil, Thickness: 0.25 mm (0.01 in)
Rhenium
Purity: 99.97% CAS No.: 7440-15-5 Physical Form: ≈3.29g/25x25mm Odour: Odourless MDL No.: MFCD00011195
Formule: Re
MW: 186.21 g/mol
Kookpunt: 5630
Smeltpunt: 3180
Dichtheid: 21.02 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011195
CAS nummer: 7440-15-5
EINECS: 231-124-5
Merck Index: 12,08343