Flessen, smalle hals, rond, LDPE, Boston-type, met schroefdop, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
312003-0001 312003-0002 312003-0004 312003-0008 312003-0016 312003-0032
NALG312003-0001EA 924 EUR
NALG312003-0001 NALG312003-0002 NALG312003-0004 NALG312003-0008 NALG312003-0016 NALG312003-0032
Flessen, smalle hals, rond, LDPE, Boston-type, met schroefdop, Nalgene®
Flessen Kunststof flessen
Boston round narrow-mouth LDPE bottles with closure, constructed from natural low-density polyethylene.
  • Good chemical resistance, heat resistant up to 80 °C
  • Well suited for trace analysis
  • More flexible and impact resistant than HDPE bottles, leakproof
  • Non sterile
Ideaal voor het opslaan en verzenden van vloeistoffen.
Uniform walls that resist splitting and puncturing. Leakproof when used with Nalgene closures.
Certificaten: Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.