Ultrafiltration membrane filters, Hydrosart®
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14429-47-D 14439-47-D 14459-47-D 14429-25-D 14439-25-D 14459-25-D 14429-44-D 14429-63-D 14439-63-D 14459-63-D 14429-76-D 14439-76-D 14459-76-D
SART14429-47-DEA 165 EUR
SART14429-47-D SART14439-47-D SART14459-47-D SART14429-25-D SART14439-25-D SART14459-25-D SART14429-44-D SART14429-63-D SART14439-63-D SART14459-63-D SART14429-76-D SART14439-76-D SART14459-76-D
Ultrafiltration membrane filters, Hydrosart®
Filters Membraanfilters
These membranes are highly hydrophilic and are often preferred for their high protein recovery when processing very dilute solutions.
  • Resistance to autoclaving
  • Ease of cleaning and extended chemical resistance