Koudelichtbronnen, KL 1600 LED en KL 2500 LED
Leverancier: SCHOTT AG Lighting and imaging
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
250400UK 250400EU 250400CH 150600 150600-SETEU
631-1032EA 1100 EUR
631-1032 631-1031 631-1030 631-1037 SCOC150600-SETEU
Koudelichtbronnen, KL 1600 LED en KL 2500 LED
Lampen Koudlichtbronnen
Deze LED koudlichtbronnen voor milieuvriendelijke PURAVIS® glasvezellichtgeleiders met rimpelvrije verlichting, daglicht zonder verandering in de kleurtemperatuur bij het dimmen worden gecombineerd met de voordelen van vezeloptica, zijnde hoge helderheid en zuivere kleuren.
  • Ecological lead-free fibre optics light guides for more visible and purer images
  • Filter slider with two positions - filter remains in the light source even when not in use
  • Compact housing with built-in handle and iron ballast for stability when used with gooseneck light guides
  • Long lifetime of at least 50000 H
KL1600 LED: Standard cold light source, with a brightness of 680 lm, is equivalent to a 150 W halogen lamp. Due to its slim design, the KL 1600 LED can be placed close to a microscope. With continuous dimming.
KL2500 LED: Advanced cold light source, with a brightness of 1100 lm, is equivalent to a 250 W halogen lamp. Controllable via USB. Intensity control with extra fine dimming at three sensitivities (fine, standard and coarse tuning). Electronic shutter function. LCD indicates brightness setting, operation mode, or error code. Powerful illumination system for the requirements of state-of-the-art, high-end stereo microscopes.
Bestelinformatie: Geleverd zonder lichtgeleider. Netsnoer moet afzonderlijk besteld worden.