Xyleen ≥97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
L13317.AP L13317.AU L13317.0F
L13317.APEA 25.1 EUR
L13317.AP L13317.AU L13317.0F
Xyleen ≥97%
Xyleen
Purity: 97+% CAS No.: 1330-20-7 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 29°(84°F) Refractive Index: 1.4970 Odour: Aromatic Melting Point: -25° Boiling Point: 137-144° Beilstein Reference: 1901563 Merck Reference: 14,10081 UN No.: 1307 MDL No.: MFCD00077264
Formule: C₆H₄(CH₃)₂
Kookpunt: 137…144 °C
Smeltpunt: < –25 °C
Dichtheid: 0,860
Vlampunt: 29 °C (84 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00077264
CAS nummer: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,II
Merck Index: 14,10081
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM
Order Now
Specificaties
Environmentally Preferable