Dichloromethane-[D₂] 99.5 atom % D for NMR spectroscopy
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Dichloromethane-d2, Methylene chloride-D2, Dichloromethane-[²H₂], Dideuteromethylene chloride, Methylene chloride-d2
176110050 176110250
ACRO176110050EA 126 EUR
ACRO176110050 ACRO176110250
Dichloromethane-[D₂] 99.5 atom % D for NMR spectroscopy
Dichloromethane-[D₂]
Formule: CD₂Cl₂
MW: 86.92 g/mol
Kookpunt: 40
Smeltpunt: –97 °C
Dichtheid: 1.36 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000882
CAS nummer: 1665-00-5
EINECS: 216-776-0
UN: 1593
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.04 % (Coulometric)
D-Enrichment ≥99.4 %
Identification C Passes test
Identification D Passes test
Alkalinity Passes test
Titration ≥85 %
NaCl and Na2SO4 ≤8 %
Non-esterified alcohols ≤4 %