Chloroform-[D₁] 99.8 atom % D for NMR spectroscopy
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D
166250250 166250500 166251000 166252500 166255000
ACRO166250250EA 33.5 EUR
ACRO166250250 ACRO166250500 ACRO166251000 ACRO166252500 ACRO166255000
Chloroform-[D₁] 99.8 atom % D for NMR spectroscopy
Chloroform-[D₁]
Formule: CDCl₃
Kookpunt: 60,8 °C (1013 hPa)
Smeltpunt: –64 °C
Dichtheid: 1,5 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000827
CAS nummer: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.02 % (Coulometric)
D-Enrichment ≥99.75 %
HPLC ≥97.5 %