Tolueen-D8 99+ atom % D for NMR spectroscopy
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
166390100 166390250
ACRO166390100EA 106 EUR
ACRO166390100 ACRO166390250
Tolueen-D8 99+ atom % D for NMR spectroscopy
Tolueen-D8
Formule: C₆D₅CD₃
Kookpunt: 110 °C (1013 hPa)
Smeltpunt: –84 °C
Dichtheid: 0,94 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00044638
CAS nummer: 2037-26-5
EINECS: 218-009-5
UN: 1294
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.05 %
D-Enrichment ≥99.0 %